AC5H30533D1D Accela ChemBio Inc.

Bromo(bromomethoxy)methane

  • MDL number MFCD31630365
  • Cas Number : 4497-29-4
  • Molecular Weight : 203.86056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY231337&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY231337.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart