AC5H30533D26 Accela ChemBio Inc.

N-(2-Hydroxyethyl)ammelide

  • MDL number MFCD31630372
  • Cas Number : 61799-46-0
  • Molecular Weight : 172.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232257&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232257.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart