AC5H30533D29 Accela ChemBio Inc.

6-(1-Piperazinyl)-1,3,5-triazine-2,4(1H,3H)-dione

  • MDL number MFCD30603738
  • Cas Number : 144742-32-5
  • Molecular Weight : 197.19454

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232271&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232271.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart