AC5H30533D2A Accela ChemBio Inc.

2-[[4-[[[4-(tert-Butyl)phenyl]sulfonyl]imino]-1-oxo-1,4-dihydro-2-naphthyl]thio]acetic Acid

  • MDL number MFCD05803546
  • Cas Number : 881487-77-0
  • Molecular Weight : 443.53584

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232308&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232308.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart