AC5H30533D32 Accela ChemBio Inc.

[4,4'-Bipyridine]-2,2'-dicarboxylic Acid

  • MDL number MFCD31630378
  • Cas Number : 85531-49-3
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232918&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232918.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart