AC5H30533D3A Accela ChemBio Inc.

3,4-Dichlorophthalic Acid

  • MDL number MFCD18973988
  • Cas Number : 56962-06-2
  • Molecular Weight : 235.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232952&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232952.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart