AC5H30533D44 Accela ChemBio Inc.

4-Chloro-2-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD23705573
  • Cas Number : 184766-53-8
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233058&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233058.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart