AC5H30533D46 Accela ChemBio Inc.

8,9-Difluorobenzo[5,6]thiepino[2,3-b]pyridin-5(10H)-one

  • MDL number MFCD31630388
  • Molecular Weight : 263.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233061&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233061.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart