AC5H30533D51 Accela ChemBio Inc.

3-Fluoroazetidine-1-ethanamine

  • MDL number MFCD31653104
  • Cas Number : 2045189-35-1
  • Molecular Weight : 118.1526432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233159&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233159.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart