AC5H30533D55 Accela ChemBio Inc.

1,1,1-Trifluoro-3-[(4-methoxybenzyl)oxy]-2-propanol

  • MDL number MFCD31630416
  • Molecular Weight : 250.2143296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233348&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233348.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart