AC5H30533D56 Accela ChemBio Inc.

N,N'-(3-Fluoro-4-methyl-8-oxo-5,6,7,8-tetrahydronaphthalene-1,7-diyl)diacetamide

  • MDL number MFCD31657913
  • Cas Number : 143655-70-3
  • Molecular Weight : 292.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233350&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233350.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart