AC5H30533D58 Accela ChemBio Inc.

2-(Chloromethyl)-5-(trifluoromethyl)pyridine Hydrochloride

  • MDL number MFCD23135652
  • Cas Number : 856250-59-4
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233372&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233372.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart