AC5H30533D5C Accela ChemBio Inc.

2-Bromo-5-(2,2,2-trifluoroethyl)pyridine

  • MDL number MFCD22381771
  • Cas Number : 1346531-45-0
  • Molecular Weight : 240.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid, or colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233387&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233387.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart