AC5H30533D5F Accela ChemBio Inc.

Methyl (S)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylate

  • MDL number MFCD31630429
  • Cas Number : 1493675-45-8
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233421&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233421.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart