AC5H30533D61 Accela ChemBio Inc.

3-Chlorofuran-2-acetic Acid

  • MDL number MFCD20660045
  • Cas Number : 1378768-90-1
  • Molecular Weight : 160.5551

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233430&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233430.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart