AC5H30533D66 Accela ChemBio Inc.

Methyl 5,6-Dimethoxybenzothiophene-2-carboxylate

  • MDL number MFCD11044560
  • Cas Number : 35212-99-8
  • Molecular Weight : 252.28628

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233497&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233497.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart