AC5H30533D69 Accela ChemBio Inc.

4-Aminophenyl Acetate

  • MDL number MFCD08691414
  • Cas Number : 13871-68-6
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233693&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233693.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart