AC5H30533D6D Accela ChemBio Inc.

6-Bromo-1,5-dimethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD30145480
  • Cas Number : 1609259-61-1
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY234121&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY234121.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart