AC5H30533D72 Accela ChemBio Inc.

2-(3,5-Dimethyl-4-pyrazolyl)benzoic Acid

  • MDL number MFCD02682016
  • Cas Number : 321309-43-7
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235434&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235434.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart