AC5H30533D74 Accela ChemBio Inc.

1,3,5-Tri(4-pyrazolyl)benzene

  • MDL number MFCD30741245
  • Cas Number : 1325728-09-3
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235439&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235439.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart