AC5H30533D75 Accela ChemBio Inc.

Pyrazolo[1,5-a]pyridine-2-acetic Acid

  • MDL number MFCD20644913
  • Cas Number : 1279821-25-8
  • Molecular Weight : 176.17202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235443&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235443.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart