AC5H30533D77 Accela ChemBio Inc.

5-(4-Methyl-1-piperazinyl)pyrazine-2-methanamine

  • MDL number MFCD20283482
  • Cas Number : 1211592-54-9
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235451&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235451.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart