AC5H30533D78 Accela ChemBio Inc.

1-Benzyl-5-phenyl-1H-pyrazole-4-carboxylic Acid

  • MDL number MFCD14462017
  • Cas Number : 905590-04-7
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235469&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235469.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart