AC5H30533D79 Accela ChemBio Inc.

1-Benzyl-5-(cyclopropylmethyl)-1H-pyrazole-4-carboxylic Acid

  • MDL number MFCD31653107
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235470&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235470.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart