AC5H30533D7B Accela ChemBio Inc.

Methyl 2-Oxo-2-(2-methylphenyl)acetate

  • MDL number MFCD00966053
  • Cas Number : 34966-54-6
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235497&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235497.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart