AC5H30533D7C Accela ChemBio Inc.

N-Boc-1-[3-(Methylthio)-1,2,4-triazin-6-yl]methylamine

  • MDL number MFCD31653108
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235506&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235506.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart