AC5H30533D80 Accela ChemBio Inc.

5-Amino-8-fluoro-1-methyl-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD31699888
  • Molecular Weight : 194.2055432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235794&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235794.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart