AC5H30533D87 Accela ChemBio Inc.

Methyl 4-Chloro-2-methoxy-5-methylbenzoate

  • MDL number MFCD31695764
  • Cas Number : 1368228-61-8
  • Molecular Weight : 214.64554

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235996&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235996.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart