AC5H30533D8E Accela ChemBio Inc.

N2-Boc-2,3-dimethyl-2,3-butanediamine

  • MDL number MFCD24617734
  • Cas Number : 1306610-31-0
  • Molecular Weight : 216.32046

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236570&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236570.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart