AC5H30533D8F Accela ChemBio Inc.

Methyl cis-4-Hydroxypyrrolidine-3-carboxylate Hydrochloride

  • MDL number MFCD22124535
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236575&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236575.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart