AC5H30533D91 Accela ChemBio Inc.

Methyl (S)-2-Amino-3-(3-bromophenyl)propanoate Hydrochloride

  • MDL number MFCD12911124
  • Cas Number : 880347-43-3
  • Molecular Weight : 294.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236621&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236621.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart