AC5H30533D96 Accela ChemBio Inc.

2,4-Dinitrophenethyl Alcohol

  • MDL number MFCD09991672
  • Cas Number : 4836-69-5
  • Molecular Weight : 212.15952

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236726&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart