AC5H30533D97 Accela ChemBio Inc.

3-Fluoro-N-(4-methoxybenzyl)-N-methyl-2-nitroaniline

  • MDL number MFCD31657964
  • Cas Number : 1458569-16-8
  • Molecular Weight : 290.2896032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236773&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236773.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart