AC5H30533D99 Accela ChemBio Inc.

2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde

  • MDL number MFCD26843559
  • Cas Number : 1192569-90-6
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236846&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236846.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart