AC5H30533D9C Accela ChemBio Inc.

6-Cyclopropylpyridine-3-methanamine

  • MDL number MFCD18255577
  • Cas Number : 1256823-20-7
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236882&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236882.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart