AC5H30533D9D Accela ChemBio Inc.

1-(Benzyloxy)-3-fluoro-2-nitrobenzene

  • MDL number MFCD14582899
  • Cas Number : 920284-79-3
  • Molecular Weight : 247.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236883&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236883.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart