AC5H30533D9E Accela ChemBio Inc.

4-(Boc-amino)-2-fluoro-5-nitrobenzonitrile

  • MDL number MFCD31695792
  • Cas Number : 473537-38-1
  • Molecular Weight : 281.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236886&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236886.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart