AC5H30533DAE Accela ChemBio Inc.

4'-Amino-3'-iodoacetophenone

  • MDL number MFCD22124432
  • Cas Number : 97776-06-2
  • Molecular Weight : 261.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237308&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237308.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart