AC5H30533DAF Accela ChemBio Inc.

Ethyl 3-(2-Methoxy-4-methylphenyl)-3-oxopropanoate

  • MDL number MFCD31695761
  • Cas Number : 125041-96-5
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237310&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237310.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart