AC5H30533DB2 Accela ChemBio Inc.

5-Benzyl-2-bromopyridine

  • MDL number MFCD22394197
  • Cas Number : 1391737-98-6
  • Molecular Weight : 248.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237402&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237402.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart