AC5H30533DB4 Accela ChemBio Inc.

6-Fluoroindoline-5-carbonitrile

  • MDL number MFCD20259150
  • Cas Number : 1702230-01-0
  • Molecular Weight : 162.1636832

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237417&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237417.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart