AC5H30533DB7 Accela ChemBio Inc.

1,2-Dibromo-3,4-dichlorobenzene

  • MDL number MFCD28783262
  • Cas Number : 100191-48-8
  • Molecular Weight : 304.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237507&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237507.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart