AC5H30533DBA Accela ChemBio Inc.

3-(5-Methyl-3-indolyl)-1-propanamine

  • MDL number MFCD03004841
  • Cas Number : 586336-25-6
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237527&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237527.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart