AC5H30533DBD Accela ChemBio Inc.

4-(Bromomethyl)azobenzene

  • MDL number MFCD18974165
  • Cas Number : 57340-21-3
  • Molecular Weight : 275.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237645&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237645.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart