AC5H30533DC4 Accela ChemBio Inc.

4-(Trifluoromethyl)furan-2(5H)-one

  • MDL number MFCD23381704
  • Cas Number : 102854-63-7
  • Molecular Weight : 152.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237799&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237799.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart