AC5H30533DC5 Accela ChemBio Inc.

4-Hydroxy-3-methoxyphenyl Acetate

  • MDL number MFCD00043558
  • Cas Number : 57244-88-9
  • Molecular Weight : 182.1733

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237801&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237801.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart