AC5H30533DC9 Accela ChemBio Inc.

(Z)-1,3-Diiodo-2-butene

  • MDL number MFCD08752489
  • Cas Number : 134005-20-2
  • Molecular Weight : 307.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237854&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237854.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart