AC5H30533DCC Accela ChemBio Inc.

N-Methyl-L-cysteine Hydrochloride

  • MDL number MFCD16876405
  • Cas Number : 14344-46-8
  • Molecular Weight : 171.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237929&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237929.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart