AC5H30533DD1 Accela ChemBio Inc.

N-(4-Aminobenzyl)biguanide

  • MDL number MFCD31699919
  • Molecular Weight : 206.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238061&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238061.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart