AC5H30533DD3 Accela ChemBio Inc.

3,4-Dichloro-5-phenylfuran-2(5H)-one

  • MDL number MFCD00728660
  • Cas Number : 72857-85-3
  • Molecular Weight : 229.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238074&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238074.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart